siRNA Details
Gene Symbol: Cyp2c39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010003.2 | NP_034133.2 | 7 | 1028 | ||
GenBank mRNA | AF047726.1 | - | 7 | 979 | ||
BC057911.1 | - | 7 | 989 | |||
BC139290.1 | - | 7 | 1028 | 8862312 | ||
BC139291.1 | - | 7 | 1028 | 8862313 |
