siRNA Details
| Gene Symbol: Dnase1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010061.5 | NP_034191.3 | 6, 7 | 849 | ||
| GenBank mRNA | AK157889.1 | - | 773 | |||
| AK158070.1 | - | 2 | 1850 | |||
| BC014718.1 | - | 6, 7 | 840 | 4925690 | ||
| BC030394.1 | - | 6, 7 | 866 | |||
| BC115570.1 | - | 6, 7 | 638 | 40102105 | ||
| BC115571.1 | - | 6, 7 | 638 | 40102106 | ||
| D83038.1 | - | 7, 8 | 719 | |||
| U00478.1 | - | 1 | 1783 | |||

Dnase1
Genomic Map