siRNA Details
| Gene Symbol: Dnase1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010061.5 | NP_034191.3 | 7 | 974 | ||
| GenBank mRNA | AK157889.1 | - | 898 | |||
| AK158070.1 | - | 2 | 1975 | |||
| BC014718.1 | - | 7 | 965 | 4925690 | ||
| BC030394.1 | - | 7 | 991 | |||
| BC115570.1 | - | 7 | 763 | 40102105 | ||
| BC115571.1 | - | 7 | 763 | 40102106 | ||
| BX638483.1 | - | 2 | 235 | |||
| D83038.1 | - | 8 | 844 | |||
| U00478.1 | - | 1 | 1908 | |||

Dnase1
Genomic Map