siRNA Details
| Gene Symbol: Egr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289925.1 | NP_001276854.1 | 3 | 259 | ||
| NM_001289927.1 | NP_001276856.1 | 2 | 133 | |||
| NM_018781.3 | NP_061251.1 | 2 | 206 | |||
| GenBank mRNA | AB221656.1 | - | 2 | 206 | ||
| AF132128.1 | - | 2 | 206 | |||
| AK141603.1 | - | 1 | 210 | |||
| BC100734.1 | - | 2 | 205 | |||
| BC103568.1 | - | 3 | 257 | 40045218 | ||
| BC103569.1 | - | 2 | 201 | |||
| BC103570.1 | - | 3 | 258 | 40045221 | ||
| BY238697.1 | - | 2 | 134 | |||
| S40835.1 | - | 2 | 114 | |||

Egr3
Genomic Map