siRNA Details
| Gene Symbol: Fcgrt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010189.3 | NP_034319.2 | 4 | 858 | ||
| GenBank mRNA | AK146933.1 | - | 4 | 823 | ||
| AK152879.1 | - | 4 | 794 | |||
| AK180969.1 | - | 1 | 47 | |||
| BC003786.1 | - | 4 | 793 | 3485649 | ||
| D37874.1 | - | 4 | 791 | |||
| L17022.1 | - | 3 | 497 | |||

Fcgrt
Genomic Map