siRNA Details
| Gene Symbol: Fgf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008002.4 | NP_032028.1 | 2 | 1046 | ||
| GenBank mRNA | AK034742.1 | - | 2 | 612 | ||
| AK045267.1 | - | 2 | 1047 | |||
| AK085244.1 | - | 2 | 368 | |||
| AK141633.1 | - | 2 | 368 | |||
| BC048229.1 | - | 2 | 711 | 6313081 | ||
| D89080.1 | - | 2 | 361 | |||
| U94517.1 | - | 2 | 361 | |||

Fgf10
Genomic Map