siRNA Details
| Gene Symbol: Fut2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271993.1 | NP_001258922.1 | 3 | 1038 | ||
| NM_018876.4 | NP_061364.2 | 2 | 934 | |||
| GenBank mRNA | AF064792.1 | - | 1 | 735 | ||
| AK033520.1 | - | 2 | 849 | |||
| AK033586.1 | - | 2 | 937 | |||
| BC138534.1 | - | 2 | 788 | 8861554 | ||
| BC145831.1 | - | 2 | 788 | 40131134 | ||

Fut2
Genomic Map