siRNA Details
| Gene Symbol: Gata4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008092.3 | NP_032118.2 | 7 | 1801 | ||
| GenBank mRNA | AB075549.1 | - | 7 | 1819 | ||
| AF179424.1 | - | 6 | 1222 | |||
| AK134639.1 | - | 6 | 825 | |||
| BC137824.1 | - | 6 | 1554 | 8860843 | ||
| BG088871.2 | - | 2 | 108 | |||

Gata4
Genomic Map