siRNA Details
| Gene Symbol: Gm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010299.3 | NP_034429.1 | 2 | 369 | ||
| GenBank mRNA | AK149642.1 | - | 2 | 261 | ||
| AK152256.1 | - | 2 | 261 | |||
| AK181597.1 | - | 2 | 177 | |||
| BC004651.1 | - | 2 | 237 | 3482848 | ||
| CT010289.1 | - | 2 | 190 | |||
| L19526.1 | - | 2 | 203 | |||
| U09816.1 | - | 2 | 246 | |||

Gm2a
Genomic Map