siRNA Details
| Gene Symbol: Gng3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010316.3 | NP_034446.1 | 2, 3 | 350 | ||
| GenBank mRNA | AK004459.1 | - | 2, 3 | 315 | ||
| AK032646.1 | - | 2, 3 | 298 | |||
| BC029680.1 | - | 2, 3 | 234 | 5365311 | ||
| U38497.1 | - | 1, 2 | 57 | |||

Gng3
Genomic Map