siRNA Details
| Gene Symbol: Emg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013536.2 | NP_038564.1 | 4, 5 | 596 | ||
| GenBank mRNA | AK081757.1 | - | 4, 5 | 574 | ||
| AK218607.1 | - | 2, 3 | 117 | |||
| BC002004.1 | - | 4, 5 | 516 | 3487480 | ||
| CB948801.1 | - | 4, 5 | 599 | |||

Emg1
Genomic Map