siRNA Details
| Gene Symbol: Hdac5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077696.1 | NP_001071164.1 | 23, 24 | 3280 | ||
| NM_001284248.1 | NP_001271177.1 | 25, 26 | 3792 | |||
| NM_001284249.1 | NP_001271178.1 | 24, 25 | 3389 | |||
| NM_001284250.1 | NP_001271179.1 | 23, 24 | 3277 | |||
| NM_010412.3 | NP_034542.3 | 23, 24 | 3277 | |||
| GenBank mRNA | AF006602.2 | - | 22, 23 | 2990 | ||
| AF207748.1 | - | 22, 23 | 2909 | |||
| AK122325.2 | - | 18, 19 | 2319 | |||
| AK139202.1 | - | 24, 25 | 3363 | |||
| AK145972.1 | - | 23, 24 | 3243 | |||
| AK146483.1 | - | 23, 24 | 3216 | |||
| AK155335.1 | - | 23, 24 | 3251 | |||
| AK161956.1 | - | 3, 4 | 409 | |||
| BC023309.1 | - | 14, 15 | 1830 | |||
| BC049926.1 | - | 23, 24 | 3211 | |||
| BC058554.1 | - | 25, 26 | 3744 | |||
| BC060609.1 | - | 22, 23 | 3071 | 5720438 | ||

Hdac5
Genomic Map