siRNA Details
| Gene Symbol: Il18bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010531.1 | NP_034661.1 | 5 | 442 | ||
| GenBank mRNA | AB019505.1 | - | 3 | 291 | ||
| AF110802.1 | - | 3 | 1669 | |||
| AF110803.1 | - | 4 | 369 | |||
| AF122907.1 | - | 5 | 442 | |||
| AK003370.1 | - | 5 | 458 | |||
| AK008452.1 | - | 4 | 373 | |||
| AK009721.1 | - | 4 | 369 | |||
| BC018332.1 | - | 4 | 352 | 4007411 | ||
| CT010164.1 | - | 3 | 291 | |||
| CT010334.1 | - | 3 | 291 | |||

Il18bp
Genomic Map