siRNA Details
| Gene Symbol: Il17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010552.3 | NP_034682.1 | 2 | 199 | ||
| GenBank mRNA | AK040420.1 | - | 2 | 200 | ||
| BC119303.1 | - | 2 | 199 | 8734052 | ||
| BC119309.1 | - | 2 | 199 | 8734058 | ||
| U43088.1 | - | 2 | 186 | |||

Il17a
Genomic Map