siRNA Details
| Gene Symbol: Il3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010556.4 | NP_034686.2 | 5 | 422 | ||
| GenBank mRNA | A02046.1 | - | 5 | 420 | ||
| AK153634.1 | - | 5 | 423 | |||
| BC125552.1 | - | 5 | 417 | 40130067 | ||
| BC125554.1 | - | 5 | 417 | 40130069 | ||
| DQ788721.1 | - | 5 | 393 | |||
| K01668.1 | - | 5 | 420 | |||
| K01850.1 | - | 5 | 420 | |||

Il3
Genomic Map