siRNA Details
| Gene Symbol: Kcnab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010599.4 | NP_034729.3 | 3 | 726 | ||
| GenBank mRNA | AK029750.1 | - | 2 | 315 | ||
| AK031806.1 | - | 3 | 707 | |||
| AK044102.1 | - | 3 | 681 | |||
| AK046194.1 | - | 3 | 716 | |||
| AK049316.1 | - | 3 | 727 | |||
| AK083134.1 | - | 3 | 728 | |||
| BC017518.1 | - | 3 | 652 | 4507029 | ||

Kcnab3
Genomic Map