siRNA Details
| Gene Symbol: Kcnab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010599.4 | NP_034729.3 | 4 | 777 | ||
| GenBank mRNA | AK029750.1 | - | 3 | 366 | ||
| AK031806.1 | - | 4 | 758 | |||
| AK044102.1 | - | 4 | 732 | |||
| AK046194.1 | - | 4 | 767 | |||
| AK049316.1 | - | 4 | 778 | |||
| AK083134.1 | - | 4 | 779 | |||
| BC017518.1 | - | 4 | 703 | 4507029 | ||

Kcnab3
Genomic Map