siRNA Details
| Gene Symbol: Lat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010689.2 | NP_034819.1 | 6, 7 | 643 | ||
| GenBank mRNA | AF036907.1 | - | 6, 7 | 643 | ||
| AK088763.1 | - | 6, 7 | 552 | |||
| AK134003.1 | - | 6, 7 | 595 | |||
| BC013337.1 | - | 6, 7 | 620 | 4191983 | ||
| BC052340.1 | - | 6, 7 | 588 | 30077071 | ||

Lat
Genomic Map