siRNA Details
| Gene Symbol: Ripply3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133229.2 | NP_573492.2 | 2 | 232 | ||
| GenBank mRNA | AB063284.1 | - | 2 | 234 | ||
| AK041355.1 | - | 2 | 222 | |||
| AK172430.1 | - | 2 | 225 | |||
| BC060291.1 | - | 2 | 190 | 30135244 | ||
| CF170896.1 | - | 2 | 232 | |||

Ripply3
Genomic Map