siRNA Details
| Gene Symbol: Mxd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010753.2 | NP_034883.2 | 4 | 344 | ||
| GenBank mRNA | AK007824.1 | - | 4 | 311 | ||
| AK131132.1 | - | 4 | 314 | |||
| AK134169.1 | - | 3 | 325 | |||
| BC011303.1 | - | 4 | 344 | 3490469 | ||
| U32395.1 | - | 4 | 207 | |||

Mxd4
Genomic Map