siRNA Details
| Gene Symbol: Mcpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008571.1 | NP_032597.1 | 4 | 399 | ||
| GenBank mRNA | AK133521.1 | - | 4 | 400 | ||
| AY007570.1 | - | 4 | 399 | |||
| BC119248.1 | - | 4 | 392 | 8733997 | ||
| BC119250.1 | - | 4 | 392 | 8733999 | ||
| J05177.1 | - | 4 | 381 | |||

Mcpt2
Genomic Map