siRNA Details
| Gene Symbol: Mnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008612.2 | NP_032638.2 | 7, 8 | 931 | ||
| GenBank mRNA | AK008134.1 | - | 7, 8 | 896 | ||
| AK008249.1 | - | 7, 8 | 934 | |||
| AK011833.1 | - | 6, 7 | 818 | |||
| AK155859.1 | - | 7, 8 | 932 | |||
| AK157721.1 | - | 7, 8 | 918 | |||
| BC089023.1 | - | 7, 8 | 920 | 30702121 | ||
| BC115630.2 | - | 7, 8 | 923 | 40102880 | ||
| U35249.1 | - | 7, 8 | 962 | |||

Mnat1
Genomic Map