siRNA Details
| Gene Symbol: Msh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010829.2 | NP_034959.2 | 3 | 559 | ||
| GenBank mRNA | AK011426.1 | - | 3 | 543 | ||
| AK040616.1 | - | 2 | 344 | |||
| AK078467.1 | - | 3 | 510 | |||
| AK083388.1 | - | 3 | 464 | |||
| AK133907.1 | - | 3 | 552 | |||
| AK161089.1 | - | 3 | 553 | |||
| BC150759.1 | - | 3 | 478 | 9087670 | ||
| BY710130.1 | - | 3 | 541 | |||
| M24919.1 | - | 3 | 842 | |||
| M80360.1 | - | 3 | 556 | |||

Msh3
Genomic Map