siRNA Details
| Gene Symbol: Mthfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008638.2 | NP_032664.1 | 2 | 235 | ||
| GenBank mRNA | AK076019.1 | - | 2 | 188 | ||
| AK076801.1 | - | 2 | 185 | |||
| AK152249.1 | - | 2 | 227 | |||
| AK159680.1 | - | 2 | 208 | |||
| AK165840.1 | - | 2 | 227 | |||
| AK166262.1 | - | 2 | 218 | |||
| BC019511.1 | - | 2 | 222 | 4164253 | ||
| J04627.1 | - | 2 | 178 | |||

Mthfd2
Genomic Map