siRNA Details
| Gene Symbol: Nnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010924.2 | NP_035054.1 | 3 | 900 | ||
| GenBank mRNA | AK006371.1 | - | 2 | 647 | ||
| BC028757.1 | - | 3 | 816 | 1067421 | ||
| BX633297.1 | - | 1 | 54 | |||
| U86105.1 | - | 3 | 947 | |||
| U86106.1 | - | 3 | 947 | |||
| U86107.1 | - | 3 | 947 | |||
| U86108.1 | - | 3 | 947 | |||
| U89377.1 | - | 4 | 816 | |||

Nnmt
Genomic Map