siRNA Details
| Gene Symbol: Otp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011021.3 | NP_035151.1 | 2 | 540 | ||
| GenBank mRNA | Y10413.1 | - | 2 | 542 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otp| Gene Symbol: Otp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011021.3 | NP_035151.1 | 2 | 540 | ||
| GenBank mRNA | Y10413.1 | - | 2 | 542 | ||