siRNA Details
| Gene Symbol: Ovol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019935.3 | NP_064319.1 | 4 | 1005 | ||
| GenBank mRNA | AF134804.1 | - | 4 | 1021 | ||
| AF134805.1 | - | 4 | 987 | |||
| AK003801.1 | - | 4 | 1040 | |||
| AK028577.1 | - | 4 | 1006 | |||
| AK132478.1 | - | 4 | 1006 | |||
| AK132505.1 | - | 4 | 1044 | |||
| AK132506.1 | - | 4 | 1040 | |||
| AK144952.1 | - | 4 | 1040 | |||
| BC021411.1 | - | 4 | 1085 | 5006712 | ||

Ovol1
Genomic Map