siRNA Details
| Gene Symbol: Ovol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019935.3 | NP_064319.1 | 4 | 945 | ||
| GenBank mRNA | AF134804.1 | - | 4 | 961 | ||
| AF134805.1 | - | 4 | 927 | |||
| AK003801.1 | - | 4 | 980 | |||
| AK028577.1 | - | 4 | 946 | |||
| AK132478.1 | - | 4 | 946 | |||
| AK132505.1 | - | 4 | 984 | |||
| AK132506.1 | - | 4 | 980 | |||
| AK144952.1 | - | 4 | 980 | |||
| BC021411.1 | - | 4 | 1025 | 5006712 | ||

Ovol1
Genomic Map