siRNA Details
| Gene Symbol: Pax7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011039.2 | NP_035169.1 | 3, 4 | 506 | ||
| GenBank mRNA | AF254422.4 | - | 3, 4 | 506 | ||
| U20792.1 | - | 2, 3 | 347 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pax7| Gene Symbol: Pax7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011039.2 | NP_035169.1 | 3, 4 | 506 | ||
| GenBank mRNA | AF254422.4 | - | 3, 4 | 506 | ||
| U20792.1 | - | 2, 3 | 347 | |||