siRNA Details
| Gene Symbol: Rhox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008818.2 | NP_032844.2 | 2 | 304 | ||
| GenBank mRNA | AK146379.1 | - | 2 | 329 | ||
| AK146393.1 | - | 2 | 328 | |||
| AK146449.1 | - | 2 | 310 | |||
| BC056204.1 | - | 3 | 312 | 30022528 | ||
| DQ058642.1 | - | 2 | 288 | |||
| M32484.1 | - | 2 | 288 | |||

Rhox5
Genomic Map