siRNA Details
| Gene Symbol: Polg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017462.2 | NP_059490.2 | 3 | 1145 | ||
| GenBank mRNA | AB121698.1 | - | 2 | 838 | ||
| AK133584.1 | - | 3 | 1071 | |||
| AK148091.1 | - | 3 | 1107 | |||
| BC053024.1 | - | 3 | 1073 | |||
| BC061472.1 | - | 3 | 1059 | 6827238 | ||
| BC080291.1 | - | 3 | 1052 | |||
| U53584.1 | - | 3 | 1065 | |||

Polg
Genomic Map