siRNA Details
| Gene Symbol: Polg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017462.2 | NP_059490.2 | 3, 4 | 1171 | ||
| GenBank mRNA | AB121698.1 | - | 2, 3 | 864 | ||
| AK133584.1 | - | 3, 4 | 1097 | |||
| AK148091.1 | - | 3, 4 | 1133 | |||
| BC053024.1 | - | 3, 4 | 1099 | |||
| BC061472.1 | - | 3, 4 | 1085 | 6827238 | ||
| BC080291.1 | - | 3, 4 | 1078 | |||
| U53584.1 | - | 3, 4 | 1091 | |||

Polg
Genomic Map