siRNA Details
| Gene Symbol: Proc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042767.2 | NP_001036232.1 | 7 | 670 | ||
| NM_001042768.2 | NP_001036233.2 | 8 | 750 | |||
| NM_008934.3 | NP_032960.3 | 7 | 682 | |||
| GenBank mRNA | AF318182.1 | - | 7 | 659 | ||
| BC013896.1 | - | 7 | 687 | 4211329 | ||
| CA468822.1 | - | 7 | 694 | |||
| CF550816.1 | - | 3 | 246 | |||
| D10445.1 | - | 6 | 598 | |||
| DV662175.1 | - | 6 | 584 | |||

Proc
Genomic Map