siRNA Details
| Gene Symbol: Rspo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138683.2 | NP_619624.2 | 4, 5 | 917 | ||
| GenBank mRNA | AB016768.1 | - | 4, 5 | 917 | ||
| AK132277.1 | - | 4, 5 | 864 | |||
| BC138831.1 | - | 3, 4 | 669 | 8861853 | ||
| BC138833.1 | - | 3, 4 | 669 | 8861855 | ||

Rspo1
Genomic Map