siRNA Details
| Gene Symbol: Rab4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029391.2 | NP_083667.1 | 1, 2 | 158 | ||
| GenBank mRNA | AB232592.1 | - | 1, 2 | 17 | ||
| AF408432.1 | - | 1, 2 | 130 | |||
| AK005314.1 | - | 1, 2 | 158 | |||
| AK140513.1 | - | 1, 2 | 91 | |||
| AK170552.1 | - | 1, 2 | 172 | |||
| BC007147.1 | - | 1, 2 | 102 | 3600193 | ||
| BC096654.1 | - | 1, 2 | 141 | 5056766 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_017355, XM_008759181, XM_008759182, X78605, FQ227674.

Rab4b
Genomic Map