siRNA Details
| Gene Symbol: Rab4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029391.2 | NP_083667.1 | 2 | 220 | ||
| GenBank mRNA | AB232592.1 | - | 2 | 79 | ||
| AF408432.1 | - | 2 | 192 | |||
| AK005314.1 | - | 2 | 220 | |||
| AK140513.1 | - | 2 | 153 | |||
| AK170552.1 | - | 2 | 234 | |||
| BC007147.1 | - | 2 | 164 | 3600193 | ||
| BC096654.1 | - | 2 | 203 | 5056766 | ||

Rab4b
Genomic Map