siRNA Details
| Gene Symbol: Nkx1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009123.2 | NP_033149.1 | 1 | 158 | ||
| GenBank mRNA | AF222444.1 | - | 1 | 158 | ||
| AF222445.1 | - | 1 | 159 | |||
| AF223361.1 | - | 1 | 160 | |||
| AF223362.1 | - | 1 | 159 | |||
| AF223363.1 | - | 1 | 158 | |||
| AF223674.1 | - | 1 | 160 | |||
| AF223675.1 | - | 1 | 160 | |||
| BC139757.1 | - | 1 | 87 | 40047346 | ||
| X75384.1 | - | 1 | 263 | |||

Nkx1-2
Genomic Map