siRNA Details
| Gene Symbol: Serpinb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009126.3 | NP_033152.3 | 7 | 1076 | ||
| GenBank mRNA | AY144683.1 | - | 7 | 1060 | ||
| AY144684.1 | - | 7 | 1060 | |||
| BC063756.1 | - | 7 | 1076 | 30288465 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinb3a| Gene Symbol: Serpinb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009126.3 | NP_033152.3 | 7 | 1076 | ||
| GenBank mRNA | AY144683.1 | - | 7 | 1060 | ||
| AY144684.1 | - | 7 | 1060 | |||
| BC063756.1 | - | 7 | 1076 | 30288465 | ||