siRNA Details
| Gene Symbol: Serpinb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011459.4 | NP_035589.1 | 7 | 1139 | ||
| GenBank mRNA | AK032659.1 | - | 7 | 1138 | ||
| AK036811.1 | - | 7 | 1075 | |||
| AK148413.1 | - | 7 | 1137 | |||
| BC011076.1 | - | 7 | 1119 | |||
| BC025121.1 | - | 7 | 1129 | 4487039 | ||
| U96703.2 | - | 7 | 1130 | |||

Serpinb8
Genomic Map