siRNA Details
| Gene Symbol: Chmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085498.2 | NP_001078967.1 | 3, 4 | 331 | ||
| GenBank mRNA | AK172123.1 | - | 3, 4 | 298 | ||
| AK219882.1 | - | 1, 2 | 59 | |||
| BB870252.1 | - | 3, 4 | 332 | |||
| BC147754.1 | - | 3, 4 | 326 | 9056703 | ||
| BC147755.1 | - | 3, 4 | 326 | 9056704 | ||
| BG078687.2 | - | 3, 4 | 246 | |||

Chmp6
Genomic Map