siRNA Details
| Gene Symbol: Chmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085498.2 | NP_001078967.1 | 3 | 285 | ||
| GenBank mRNA | AK172123.1 | - | 3 | 252 | ||
| AK219882.1 | - | 1 | 13 | |||
| BB870252.1 | - | 3 | 286 | |||
| BC147754.1 | - | 3 | 280 | 9056703 | ||
| BC147755.1 | - | 3 | 280 | 9056704 | ||
| BG078687.2 | - | 3 | 200 | |||

Chmp6
Genomic Map