siRNA Details
| Gene Symbol: Gm609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005854.2 | NP_001005854.2 | 2 | 397 | ||
| GenBank mRNA | AB853320.1 | - | 2 | 413 | ||
| AK136536.1 | - | 2 | 415 | |||
| BC054854.1 | - | 2 | 396 | 6529072 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm609| Gene Symbol: Gm609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005854.2 | NP_001005854.2 | 2 | 397 | ||
| GenBank mRNA | AB853320.1 | - | 2 | 413 | ||
| AK136536.1 | - | 2 | 415 | |||
| BC054854.1 | - | 2 | 396 | 6529072 | ||