siRNA Details
| Gene Symbol: Surf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013678.2 | NP_038706.1 | 6 | 729 | ||
| GenBank mRNA | AK004931.1 | - | 2 | 2802 | ||
| AK157125.1 | - | 6 | 743 | |||
| AK165559.1 | - | 6 | 736 | |||
| AK172018.1 | - | 6 | 729 | |||
| BC049779.1 | - | 5 | 2060 | |||
| BC096401.1 | - | 6 | 729 | 1365532 | ||

Surf2
Genomic Map