siRNA Details
| Gene Symbol: Svs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009301.2 | NP_033327.1 | 2 | 128 | ||
| GenBank mRNA | AK007282.1 | - | 2 | 129 | ||
| BC114349.1 | - | 2 | 132 | 40053311 | ||
| S60687.1 | - | 1 | 18 | |||
| X57139.1 | - | 2 | 156 | |||
| X63161.1 | - | 2 | 108 | |||

Svs5
Genomic Map