siRNA Details
Gene Symbol: Hsh2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197944.1 | NP_922935.1 | 5 | 504 | ||
GenBank mRNA | AK157000.1 | - | 537 | |||
AY319653.1 | - | 5 | 504 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hsh2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197944.1 | NP_922935.1 | 5 | 504 | ||
GenBank mRNA | AK157000.1 | - | 537 | |||
AY319653.1 | - | 5 | 504 |