siRNA Details
| Gene Symbol: Lrrc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153074.3 | NP_694714.1 | 2 | 337 | ||
| GenBank mRNA | AJ422149.1 | - | 1 | 154 | ||
| AK080022.1 | - | 2 | 338 | |||
| AK171924.1 | - | 2 | 327 | |||
| BC137696.1 | - | 2 | 297 | 8860715 | ||

Lrrc25
Genomic Map