siRNA Details
| Gene Symbol: Mfsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114662.1 | NP_001108134.1 | 2 | 492 | ||
| NM_172510.4 | NP_766098.2 | 2 | 492 | |||
| GenBank mRNA | AK038882.1 | - | 2 | 385 | ||
| AK044667.1 | - | 2 | 365 | |||
| AK085484.1 | - | 2 | 364 | |||
| AK162495.1 | - | 2 | 396 | |||
| BC058790.1 | - | 2 | 341 | 6395070 | ||

Mfsd4
Genomic Map