siRNA Details
| Gene Symbol: Tnrc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144812.2 | NP_659061.2 | 23 | 5163 | ||
| NM_177124.4 | NP_796098.3 | 20 | 4954 | |||
| GenBank mRNA | AK051174.1 | - | 5 | 551 | ||
| AK052247.1 | - | 15 | 1917 | |||
| AK122440.1 | - | 9 | 1155 | |||
| AK162702.1 | - | 14 | 1654 | |||
| AK162788.1 | - | 15 | 2213 | |||
| BC017531.1 | - | 2 | 657 | |||
| BC080750.1 | - | 4 | 378 | |||
| BC117548.1 | - | 2 | 415 | |||
| BC118054.1 | - | 2 | 415 | |||

Tnrc6b
Genomic Map