siRNA Details
| Gene Symbol: Tnrc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144812.2 | NP_659061.2 | 24 | 5214 | ||
| NM_177124.4 | NP_796098.3 | 21 | 5005 | |||
| GenBank mRNA | AK051174.1 | - | 6 | 602 | ||
| AK052247.1 | - | 16 | 1968 | |||
| AK122440.1 | - | 10 | 1206 | |||
| AK162702.1 | - | 15 | 1705 | |||
| AK162788.1 | - | 16 | 2264 | |||
| BC017531.1 | - | 3 | 708 | |||
| BC080750.1 | - | 5 | 429 | |||
| BC117548.1 | - | 3 | 466 | |||
| BC118054.1 | - | 3 | 466 | |||

Tnrc6b
Genomic Map