siRNA Details
| Gene Symbol: Tnrc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144812.2 | NP_659061.2 | 25 | 5378 | ||
| NM_177124.4 | NP_796098.3 | 22 | 5169 | |||
| GenBank mRNA | AK051174.1 | - | 7 | 766 | ||
| AK052247.1 | - | 17 | 2132 | |||
| AK122440.1 | - | 11 | 1370 | |||
| AK162702.1 | - | 16 | 1869 | |||
| AK162788.1 | - | 17 | 2428 | |||
| BC017531.1 | - | 4 | 872 | |||
| BC080750.1 | - | 6 | 593 | |||
| BC117548.1 | - | 4 | 630 | |||
| BC118054.1 | - | 4 | 630 | |||

Tnrc6b
Genomic Map